RIE156321040 Rieke Metals, Inc.

2-chloro-3-(7-chloroheptanoyl)pyridine

  • MDL number MFCD02260514
  • Cas Number : 914203-38-6
  • Molecular Weight : 260.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5541F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5541F
Availability details for
Quantity
  • Availability
  • Add to Cart