RIE156321042 Rieke Metals, Inc.

2-chloro-5-(6-chlorohexanoyl)pyridine

  • MDL number MFCD02260518
  • Cas Number : 914203-40-0
  • Molecular Weight : 246.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5542E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5542E
Availability details for
Quantity
  • Availability
  • Add to Cart