RIE156321044 Rieke Metals, Inc.

2-Chloro-3-(2-furanoyl)pyridine

  • MDL number MFCD02260525
  • Cas Number : 914203-42-2
  • Molecular Weight : 207.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5544

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5544
Availability details for
Quantity
  • Availability
  • Add to Cart