RIE156321047 Rieke Metals, Inc.

5-(2-chloronicotinoyl)-2-furoic acid

  • MDL number MFCD02260528
  • Cas Number : 914203-45-5
  • Molecular Weight : 251.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5547

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5547
Availability details for
Quantity
  • Availability
  • Add to Cart