RIE156321048 Rieke Metals, Inc.

2-bromo-5-(4-nitrobenzoyl)thiophene

  • MDL number MFCD03841024
  • Cas Number : 909421-68-7
  • Molecular Weight : 312.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-2-thienyl)(4-nitrophenyl)methanone
Inchi Code 1S/C11H6BrNO3S/c12-10-6-5-9(17-10)11(14)7-1-3-8(4-2-7)13(15)16/h1-6H
Inchi Key LHVCLCGDOGUBPC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5553

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5553
Availability details for
Quantity
  • Availability
  • Add to Cart