RIE156321049 Rieke Metals, Inc.

2-chloro-5-(4-nitrobenzoyl)thiophene

  • MDL number MFCD03841025
  • Cas Number : 909421-69-8
  • Molecular Weight : 267.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5555

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5555
Availability details for
Quantity
  • Availability
  • Add to Cart