RIE156321050 Rieke Metals, Inc.

Ethyl 5-(3-oxocyclohexyl)-2-thiophenecarboxylate

  • MDL number MFCD03841026
  • Cas Number : 909421-70-1
  • Molecular Weight : 252.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5556

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5556
Availability details for
Quantity
  • Availability
  • Add to Cart