RIE156321051 Rieke Metals, Inc.

Ethyl 5-(3-oxocyclohexyl)-2-furoate

  • MDL number MFCD03841030
  • Cas Number : 883898-17-7
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5560

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5560
Availability details for
Quantity
  • Availability
  • Add to Cart