RIE156321052 Rieke Metals, Inc.

2-Chloro-5-[(3-oxocyclohexyl)methyl]pyridine

  • MDL number MFCD03841031
  • Cas Number : 909421-71-2
  • Molecular Weight : 223.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5561

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5561
Availability details for
Quantity
  • Availability
  • Add to Cart