RIE156321054 Rieke Metals, Inc.

3-(2-methylbenzoyl)thiophene

  • MDL number MFCD07699006
  • Cas Number : 67869-14-1
  • Molecular Weight : 202.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-02
Availability details for
Quantity
  • Availability
  • Add to Cart