RIE156321055 Rieke Metals, Inc.

3-(4-methylbenzoyl)thiophene

  • MDL number MFCD07699008
  • Cas Number : 118993-65-0
  • Molecular Weight : 202.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-04

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-04
Availability details for
Quantity
  • Availability
  • Add to Cart