RIE156321056 Rieke Metals, Inc.

3-(2-methoxybenzoyl)thiophene

  • MDL number MFCD07699009
  • Cas Number : 109106-65-2
  • Molecular Weight : 218.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-05
Availability details for
Quantity
  • Availability
  • Add to Cart