RIE156321057 Rieke Metals, Inc.

3-(3-methoxybenzoyl)thiophene

  • MDL number MFCD07699010
  • Cas Number : 861223-63-4
  • Molecular Weight : 218.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-06

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-06
Availability details for
Quantity
  • Availability
  • Add to Cart