RIE156321059 Rieke Metals, Inc.

3-(2-cyanobenzoyl)thiophene

  • MDL number MFCD07699012
  • Cas Number : 909421-73-4
  • Molecular Weight : 213.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-08
Availability details for
Quantity
  • Availability
  • Add to Cart