RIE156321062 Rieke Metals, Inc.

3-(2-carboethoxybenzoyl)thiophene

  • MDL number MFCD07699015
  • Cas Number : 904633-30-3
  • Molecular Weight : 260.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-(3-thienylcarbonyl)benzoate
Inchi Code 1S/C14H12O3S/c1-2-17-14(16)12-6-4-3-5-11(12)13(15)10-7-8-18-9-10/h3-9H,2H2,1H3
Inchi Key RNDFBYVEETWXBT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5564-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-11
Availability details for
Quantity
  • Availability
  • Add to Cart