RIE156321063 Rieke Metals, Inc.

3-(3-carboethoxybenzoyl)thiophene

  • MDL number MFCD07699016
  • Cas Number : 896618-51-2
  • Molecular Weight : 260.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-12
Availability details for
Quantity
  • Availability
  • Add to Cart