RIE156321066 Rieke Metals, Inc.

3-(4-thiomethylbenzoyl)thiophene

  • MDL number MFCD07699019
  • Cas Number : 896618-54-5
  • Molecular Weight : 234.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-15
Availability details for
Quantity
  • Availability
  • Add to Cart