RIE156321069 Rieke Metals, Inc.

3-(3-Fluorobenzoyl)thiophene

  • MDL number MFCD07699024
  • Cas Number : 896618-57-8
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-22

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-22
Availability details for
Quantity
  • Availability
  • Add to Cart