RIE156321071 Rieke Metals, Inc.

3-(2,3-Dimethylbenzoyl)thiophene

  • MDL number MFCD07699026
  • Cas Number : 896618-58-9
  • Molecular Weight : 216.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dimethylphenyl)(3-thienyl)methanone
Inchi Code 1S/C13H12OS/c1-9-4-3-5-12(10(9)2)13(14)11-6-7-15-8-11/h3-8H,1-2H3
Inchi Key SSQQVQVYUDJNFC-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5564-26

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-26
Availability details for
Quantity
  • Availability
  • Add to Cart