RIE156321074 Rieke Metals, Inc.

3-(2,6-Dimethylbenzoyl)thiophene

  • MDL number MFCD07699029
  • Cas Number : 898771-06-7
  • Molecular Weight : 216.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-29

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-29
Availability details for
Quantity
  • Availability
  • Add to Cart