RIE156321078 Rieke Metals, Inc.

3-(4-Chloro-3-fluorobenzoyl)thiophene

  • MDL number MFCD07699033
  • Cas Number : 898771-18-1
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-33
Availability details for
Quantity
  • Availability
  • Add to Cart