RIE156321080 Rieke Metals, Inc.

3-(2-Chlorobenzoyl)thiophene

  • MDL number MFCD07699035
  • Cas Number : 898771-24-9
  • Molecular Weight : 222.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-36

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-36
Availability details for
Quantity
  • Availability
  • Add to Cart