RIE156321081 Rieke Metals, Inc.

3-(2-Fluorobenzoyl)thiophene

  • MDL number MFCD07699036
  • Cas Number : 898771-27-2
  • Molecular Weight : 206.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-37
Availability details for
Quantity
  • Availability
  • Add to Cart