RIE156321082 Rieke Metals, Inc.

3-(2-trifluoromethylbenzoyl)thiophene

  • MDL number MFCD07699037
  • Cas Number : 898771-30-7
  • Molecular Weight : 256.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-thienyl[2-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C12H7F3OS/c13-12(14,15)10-4-2-1-3-9(10)11(16)8-5-6-17-7-8/h1-7H
Inchi Key QFUBEOXTTUOKAN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5564-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-43
Availability details for
Quantity
  • Availability
  • Add to Cart