RIE156321083 Rieke Metals, Inc.

3-(3-Trifluoromethylbenzoyl)thiophene

  • MDL number MFCD07699038
  • Cas Number : 898771-33-0
  • Molecular Weight : 256.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-44
Availability details for
Quantity
  • Availability
  • Add to Cart