RIE156321086 Rieke Metals, Inc.

3-(2-Chloro-4-fluorobenzoyl)thiophene

  • MDL number MFCD07699041
  • Cas Number : 898771-41-0
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-50

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-50
Availability details for
Quantity
  • Availability
  • Add to Cart