RIE156321088 Rieke Metals, Inc.

3-(4-Chloro-2-fluorobenzoyl)thiophene

  • MDL number MFCD07699043
  • Cas Number : 898771-47-6
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-chloro-2-fluorophenyl)(3-thienyl)methanone
Inchi Code 1S/C11H6ClFOS/c12-8-1-2-9(10(13)5-8)11(14)7-3-4-15-6-7/h1-6H
Inchi Key RMBHLKZVKYMVDR-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5564-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-52
Availability details for
Quantity
  • Availability
  • Add to Cart