RIE156321089 Rieke Metals, Inc.

3-(2,3-Dichlorobenzoyl)thiophene

  • MDL number MFCD07699044
  • Cas Number : 898771-50-1
  • Molecular Weight : 257.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-53

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-53
Availability details for
Quantity
  • Availability
  • Add to Cart