RIE156321090 Rieke Metals, Inc.

3-(2,4-Dichlorobenzoyl)thiophene

  • MDL number MFCD07699045
  • Cas Number : 898771-52-3
  • Molecular Weight : 257.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-54
Availability details for
Quantity
  • Availability
  • Add to Cart