RIE156321092 Rieke Metals, Inc.

3-(3,4-Dichlorobenzoyl)thiophene

  • MDL number MFCD07699047
  • Cas Number : 898771-56-7
  • Molecular Weight : 257.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-56
Availability details for
Quantity
  • Availability
  • Add to Cart