RIE156321095 Rieke Metals, Inc.

3-(3,4-Difluorobenzoyl)thiophene

  • MDL number MFCD07699050
  • Cas Number : 898771-62-5
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-59
Availability details for
Quantity
  • Availability
  • Add to Cart