RIE156321097 Rieke Metals, Inc.

3-(3,4,5-trifluorobenzoyl)thiophene

  • MDL number MFCD07699052
  • Cas Number : 898771-66-9
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5564-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5564-61
Availability details for
Quantity
  • Availability
  • Add to Cart