RIE156321099 Rieke Metals, Inc.

Cyclobutyl 3-thienyl ketone

  • MDL number MFCD07699055
  • Cas Number : 898771-70-5
  • Molecular Weight : 166.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5565-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5565-02
Availability details for
Quantity
  • Availability
  • Add to Cart