RIE156321100 Rieke Metals, Inc.

Cyclopentyl 3-thienyl ketone

  • MDL number MFCD07699056
  • Cas Number : 898771-72-7
  • Molecular Weight : 180.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5565-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5565-03
Availability details for
Quantity
  • Availability
  • Add to Cart