RIE156321101 Rieke Metals, Inc.

Ethyl 4-oxo-4-(3-thienyl)butyrate

  • MDL number MFCD07699058
  • Cas Number : 473693-79-7
  • Molecular Weight : 212.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5565-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5565-05
Availability details for
Quantity
  • Availability
  • Add to Cart