RIE156321103 Rieke Metals, Inc.

Ethyl 7-oxo-7-(3-thienyl)heptanoate

  • MDL number MFCD07699061
  • Cas Number : 898771-76-1
  • Molecular Weight : 254.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5565-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5565-08
Availability details for
Quantity
  • Availability
  • Add to Cart