RIE156321104 Rieke Metals, Inc.

Ethyl 8-oxo-8-(3-thienyl)octanoate

  • MDL number MFCD07699062
  • Cas Number : 898771-78-3
  • Molecular Weight : 268.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5565-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5565-09
Availability details for
Quantity
  • Availability
  • Add to Cart