RIE156321105 Rieke Metals, Inc.

3-Bromo-5-(2-ethoxycarbonylbenzoyl)pyridine

  • MDL number MFCD07699063
  • Cas Number : 898771-80-7
  • Molecular Weight : 334.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5566

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5566
Availability details for
Quantity
  • Availability
  • Add to Cart