RIE156321106 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl methyl ketone

  • MDL number MFCD06209843
  • Cas Number : 773097-36-2
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01
Availability details for
Quantity
  • Availability
  • Add to Cart