RIE156321108 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl propyl ketone

  • MDL number MFCD07699065
  • Cas Number : 898771-84-1
  • Molecular Weight : 226.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01B
Availability details for
Quantity
  • Availability
  • Add to Cart