RIE156321109 Rieke Metals, Inc.

Butyl 5-(1,3-dioxolan-2-yl)-2-thienyl ketone

  • MDL number MFCD07699066
  • Cas Number : 898771-86-3
  • Molecular Weight : 240.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01C
Availability details for
Quantity
  • Availability
  • Add to Cart