RIE156321110 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl pentyl ketone

  • MDL number MFCD07699067
  • Cas Number : 898771-88-5
  • Molecular Weight : 254.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01D
Availability details for
Quantity
  • Availability
  • Add to Cart