RIE156321112 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl heptyl ketone

  • MDL number MFCD07699069
  • Cas Number : 898771-92-1
  • Molecular Weight : 282.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01F
Availability details for
Quantity
  • Availability
  • Add to Cart