RIE156321114 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl nonyl ketone

  • MDL number MFCD07699071
  • Cas Number : 898771-96-5
  • Molecular Weight : 310.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01H

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01H
Availability details for
Quantity
  • Availability
  • Add to Cart