RIE156321115 Rieke Metals, Inc.

Decyl 5-(1,3-Dioxolan-2-yl)-2-thienyl ketone

  • MDL number MFCD07699072
  • Cas Number : 898771-98-7
  • Molecular Weight : 324.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01I

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01I
Availability details for
Quantity
  • Availability
  • Add to Cart