RIE156321116 Rieke Metals, Inc.

5-(1,3-Dioxolan-2-yl)-2-thienyl undecyl ketone

  • MDL number MFCD07699073
  • Cas Number : 898772-00-4
  • Molecular Weight : 338.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-01J

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-01J
Availability details for
Quantity
  • Availability
  • Add to Cart