RIE156321118 Rieke Metals, Inc.

Methyl 5-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylate

  • MDL number MFCD07699075
  • Cas Number : 898772-05-9
  • Molecular Weight : 214.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-02
Availability details for
Quantity
  • Availability
  • Add to Cart