RIE156321119 Rieke Metals, Inc.

Ethyl 5-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylate

  • MDL number MFCD07699076
  • Cas Number : 898772-08-2
  • Molecular Weight : 228.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5567-02A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-02A
Availability details for
Quantity
  • Availability
  • Add to Cart