RIE156321120 Rieke Metals, Inc.

Propyl 5-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylate

  • MDL number MFCD07699077
  • Cas Number : 898772-11-7
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5567-02b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-02b
Availability details for
Quantity
  • Availability
  • Add to Cart