RIE156321121 Rieke Metals, Inc.

Butyl 5-(1,3-Dioxolan-2-yl)-2-thiophenecarboxylate

  • MDL number MFCD07699078
  • Cas Number : 898772-14-0
  • Molecular Weight : 256.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name butyl 5-(1,3-dioxolan-2-yl)-2-thiophenecarboxylate
Inchi Code 1S/C12H16O4S/c1-2-3-6-14-11(13)9-4-5-10(17-9)12-15-7-8-16-12/h4-5,12H,2-3,6-8H2,1H3
Inchi Key JCHFEPFEKYVYNG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5567-02c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5567-02c
Availability details for
Quantity
  • Availability
  • Add to Cart